Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAT1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$206.50 - $499.50
Specifications
| Antigen | SAT1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11041622T
![]() |
Novus Biologicals
NB11041622SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB11041622
![]() |
Novus Biologicals
NB11041622 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
SAT1 Polyclonal specifically detects SAT1 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Goat, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SAT1 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Biology, Hypoxia, Mitochondrial Markers | |
| PBS with 0.02% Sodium Azide | |
| DC21, diamine acetyltransferase 1, diamine N-acetyltransferase 1, EC 2.3.1.57, KFSD, Polyamine N-acetyltransferase 1, Putrescine acetyltransferase, SAT, Spermidine/spermine N(1)-acetyltransferase 1, spermidine/spermine N1-acetyltransferase, spermidine/spermine N1-acetyltransferase 1, spermidine/spermine N1-acetyltransferase alpha, SSAT-1, SSATKFSDX | |
| SAT1 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, rat, pig, chicken, cow, goat, and Xenopus. Immunogen sequence has 86% homology to Zebrafish. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Chicken, Goat, Xenopus species | |
| P21673 | |
| 6303 | |
| A synthetic peptide made to a C-terminal region within residues 100-171 of human SAT1. [Swiss-Prot# P21673] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 27 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title