Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAT1 Antibody (CPTC-SAT1-3), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308414F
Description
SAT1 Monoclonal antibody specifically detects SAT1 in Human samples. It is validated for ELISA, Protein ArraySpecifications
SAT1 | |
Monoclonal | |
FITC | |
DC21, diamine acetyltransferase 1, diamine N-acetyltransferase 1, EC 2.3.1.57, KFSD, Polyamine N-acetyltransferase 1, Putrescine acetyltransferase, SAT, Spermidine/spermine N(1)-acetyltransferase 1, spermidine/spermine N1-acetyltransferase, spermidine/spermine N1-acetyltransferase 1, spermidine/spermine N1-acetyltransferase alpha, SSAT-1, SSATKFSDX | |
Recombinant full-length human SAT1 protein (Uniprot: P21673) | |
100 μg | |
Cancer, Cell Biology, Hypoxia, Mitochondrial Markers | |
6303 | |
Store at 4°C in the dark. | |
IgG2a κ |
ELISA, Protein Array | |
CPTC-SAT1-3 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction