Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAT1 Mouse anti-Human, Clone: CPTC-SAT1-3, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | SAT1 |
---|---|
Clone | CPTC-SAT1-3 |
Concentration | 0.2 mg/ml |
Dilution | ELISA, Protein Array |
Applications | ELISA, Peptide Array |
Description
SAT1 Monoclonal specifically detects SAT1 in Human samples. It is validated for ELISA, Protein Array.Specifications
SAT1 | |
0.2 mg/ml | |
ELISA, Peptide Array | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Hypoxia, Mitochondrial Markers | |
10 mM PBS with 0.05% BSA | |
6303 | |
IgG2a κ | |
Protein A or G purified |
CPTC-SAT1-3 | |
ELISA, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DC21, diamine acetyltransferase 1, diamine N-acetyltransferase 1, EC 2.3.1.57, KFSD, Polyamine N-acetyltransferase 1, Putrescine acetyltransferase, SAT, Spermidine/spermine N(1)-acetyltransferase 1, spermidine/spermine N1-acetyltransferase, spermidine/spermine N1-acetyltransferase 1, spermidine/spermine N1-acetyltransferase alpha, SSAT-1, SSATKFSDX | |
Recombinant full-length human SAT1 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title