Learn More
Invitrogen™ SAT1 Polyclonal Antibody

Description
Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended. Suggested positive control: antigen standard for SAT1 (transient overexpression lysate).
Specifications
Specifications
| Antigen | SAT1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SAT1 |
| Gene Accession No. | P21673, P48026, Q28999, Q3T0Q0, Q8AXL1 |
| Gene Alias | AA617398; Ab2-402; CAON; DC21; diamine acetyltransferase 1; diamine acetyltransferase 1; spermidine/spermine N1-acetyltransferase 1; diamine N-acetyltransferase 1; diamine N-acetyltransferase 1 a; EC 2.3.1.57; EDM4; epid34; hypothetical protein LOC508861; kfsd; KFSDX; Polyamine N-acetyltransferase 1; putrescine acetyltransferase; SAT; Sat1; SAT-1; sat1.S; sat1a; sat1-a; sat1-b; Slc26a1; solute carrier family 26 (anion exchanger), member 1; solute carrier family 26 (sulfate transporter), member 1; solute carrier family 26 member 1; solute carrier family 26, member 1; Spermidine / spermine N1-acyltransferase (diamine acetyltransferase); spermidine/spermine N(1)-acetyltransferase 1; spermidine/spermine N1-acetyl transferase 1; spermidine/spermine N1-acetyltransferase; spermidine/spermine N1-acetyltransferase 1; spermidine/spermine N1-acetyltransferase 1 a; spermidine/spermine N1-acetyltransferase 1 S homeolog; spermidine/spermine N1-acetyltransferase alpha; spermidine/spermine N-acetyltransferase; SSAT; SSAT-1; sulfate anion tranporter AT1; sulfate anion transporter 1; sulfate anion transporter-1; sulfate/anion transporter SAT-1 protein; XELAEV_18014544mg |
| Gene Symbols | SAT1, sat1.S |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.