Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596145
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596145 Supplier Invitrogen™ Supplier No. PA596145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: THP-1; Cellular Location: Cytoplasm Immunogen sequence: MAKFVIRPAT AADCSDILRL IKELAKYEYM EEQVILTEKD LLEDGFGEHP FYHCLVAEVP KEHWTPEGHS IVGFAMYYFT YDPWIGKLLY LEDFFVMSDY RGFGIGSEIL KNLSQVAMRC RCSSMHFLVA EWNEPSINFY KRRGASDLSS EEGWRLFKID KEYLLKMATE E.

Spermidine/spermine-N1-acetyltransferase (SSAT1) is a short-lived polyamine catabolic enzyme which catalyzes the acetylation of polyamines. This highly regulated enzyme allows a fine attenuation of the intracellular concentration of polyamines. Induction of SSAT1 plays an important role in polyamine homoeostasis, since the N1-acetylated polyamines can be excreted or oxidized by acetylpolyamine oxidase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SAT1
Gene Accession No. P21673
Gene Alias AA617398; Ab2-402; CAON; DC21; diamine acetyltransferase 1; diamine acetyltransferase 1; spermidine/spermine N1-acetyltransferase 1; diamine N-acetyltransferase 1; diamine N-acetyltransferase 1 a; EC 2.3.1.57; EDM4; epid34; hypothetical protein LOC508861; kfsd; KFSDX; Polyamine N-acetyltransferase 1; putrescine acetyltransferase; SAT; Sat1; SAT-1; sat1.S; sat1a; sat1-a; sat1-b; Slc26a1; solute carrier family 26 (anion exchanger), member 1; solute carrier family 26 (sulfate transporter), member 1; solute carrier family 26 member 1; solute carrier family 26, member 1; Spermidine / spermine N1-acyltransferase (diamine acetyltransferase); spermidine/spermine N(1)-acetyltransferase 1; spermidine/spermine N1-acetyl transferase 1; spermidine/spermine N1-acetyltransferase; spermidine/spermine N1-acetyltransferase 1; spermidine/spermine N1-acetyltransferase 1 a; spermidine/spermine N1-acetyltransferase 1 S homeolog; spermidine/spermine N1-acetyltransferase alpha; spermidine/spermine N-acetyltransferase; SSAT; SSAT-1; sulfate anion tranporter AT1; sulfate anion transporter 1; sulfate anion transporter-1; sulfate/anion transporter SAT-1 protein; XELAEV_18014544mg
Gene Symbols SAT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-171 of human SAT1 (NP_0029611).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6303
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.