Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAT2 Monoclonal Antibody (OTI1C9)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525941

Catalog No. PIMA525941


Only null left
Add to Cart

Description

Description

SAT2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

SAT2 is an enzyme which catalyzes the acetylation of polyamines. Substrate specificity: norspermidine > spermidine = spermine >> N(1)acetylspermine = putrescine.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SAT2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96F10
Sat2
Full length human recombinant protein of SAT2 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI1C9
Unconjugated
Sat2
2610016A03Rik; Diamine acetyltransferase 2; diamine N-acetyltransferase 2; EGK_08139; Polyamine N-acetyltransferase 2; Sat2; Spermidine/spermine N(1)-acetyltransferase 2; spermidine/spermine N1-acetyl transferase 2; spermidine/spermine N1-acetyltransferase family member 2; Ssat2; SSAT-2; thialysine N-epsilon-acetyltransferase
Mouse
Affinity Chromatography
RUO
112483
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.