Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554566
Description
Immunogen sequence: IYVMPEYRGQ GIGSKIIKKV AEVALDKGCS QFRLAVLDWN QRAMDLYKAL GAQDLTEAEG WHFFCFQGEA TRKLAGK Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.
SAT2 is an enzyme which catalyzes the acetylation of polyamines. Substrate specificity: norspermidine > spermidine = spermine >> N(1)acetylspermine = putrescine.
Specifications
SAT2 | |
Polyclonal | |
Unconjugated | |
Sat2 | |
2610016A03Rik; Diamine acetyltransferase 2; diamine N-acetyltransferase 2; EGK_08139; Polyamine N-acetyltransferase 2; Sat2; Spermidine/spermine N(1)-acetyltransferase 2; spermidine/spermine N1-acetyl transferase 2; spermidine/spermine N1-acetyltransferase family member 2; Ssat2; SSAT-2; thialysine N-epsilon-acetyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
112483 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96F10 | |
Sat2 | |
Recombinant protein corresponding to Human SAT2. Recombinant protein control fragment (Product #RP-93296). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction