Learn More
SATB1 Mouse anti-Human, Clone: OTI11H4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500523

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a matrix protein which binds nuclear matrix and scaffold-associating DNAs through a unique nuclear architecture. The protein recruits chromatin-remodeling factors in order to regulate chromatin structure and gene expression. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
SATB1 | |
Monoclonal | |
Unconjugated | |
SATB1 | |
SATB homeobox 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6304 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI11H4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q01826 | |
SATB1 | |
Full-length protein expressed in 293T cell transfected with human SATB1 expression vector | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.