Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SATB1 Mouse anti-Human, Clone: OTI3E12, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500578

Description
This gene encodes a matrix protein which binds nuclear matrix and scaffold-associating DNAs through a unique nuclear architecture. The protein recruits chromatin-remodeling factors in order to regulate chromatin structure and gene expression. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
SATB1 | |
Monoclonal | |
0.93 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q01826 | |
SATB1 | |
Full length human recombit protein of human SATB1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3E12 | |
Unconjugated | |
SATB1 | |
SATB homeobox 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6304 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction