Learn More
Invitrogen™ SATB1 Polyclonal Antibody

Description
A suggested positive control is SK-N-SH cell lysate. PA5-20535 can be used with blocking peptide PEP-0655.
Specifications
Specifications
| Antigen | SATB1 |
| Applications | ChIP Assay, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Satb1 |
| Gene Accession No. | Q01826, Q60611 |
| Gene Alias | 2610306G12Rik; AW413156; DNA-binding protein SATB1; SATB homeobox 1; Satb1; special AT-rich sequence binding protein 1; special AT-rich sequence binding protein 1 (binds to nuclear matrix/scaffold-associating DNA); special AT-rich sequence-binding protein 1 |
| Gene Symbols | Satb1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.