Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SATB2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP103328JF646
Description
SATB2 Polyclonal antibody specifically detects SATB2 in Human samples. It is validated for Western BlotSpecifications
SATB2 | |
Polyclonal | |
Western Blot | |
DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2 | |
The immunogen recognized by this antibody maps to a region between residue 683 and 733 of human Special AT-rich sequence-binding protein 2 using the numbering given in entry NP_056080.1 (GeneID 23314). | |
0.1 mL | |
Signal Transduction | |
23314 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction