Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SATB2 Antibody (SATB2/4374R), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308808IR
Description
SATB2 Monoclonal antibody specifically detects SATB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SATB2 | |
Monoclonal | |
DyLight 755 | |
DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2 | |
Synthetic peptide corresponding to residues within aa200-300 corresponding to human SATB2 (Uniprot: Q9UPW6) | |
100 μg | |
Signal Transduction | |
23314 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
SATB2/4374R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction