Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SATB2 Monoclonal Antibody (CL0276)
GREENER_CHOICE

Catalog No. PIMA531339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531339 Supplier Invitrogen™ Supplier No. MA531339
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: VSQAVFARVA FNRTQGLLSE ILRKEEDPRT ASQSLLVNLR AMQNFLNLPE VERDRIYQDE RERSMNPNVS MVSSASSSPS SSRTPQAKTS TPTTDLPIKV DGANINITAA IYDEIQQEMK RAK Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

DNA-binding protein SATB2 binds to DNA at the nuclear matrix- or scaffold associated regions. It is thought to recognize the sugar-phosphate structure of double-stranded DNA. SATB2 is a transcription factor controlling nuclear gene expression by binding to matrix attachment regions (MARs) of DNA and inducing a local chromatin-loop remodeling. It also acts as a docking site for several chromatin remodeling enzymes by recruiting corepressors (HDACs) or coactivators (HATs) directly to promoters and enhancers. SATB2 is required for the initiation of the upper-layer neurons (UL1) specific genetic program and for the inactivation of deep-layer neurons (DL) and UL2 specific genes - probably by modulating BCL11B expression. It is a repressor of Ctip2 and a regulatory determinant of corticocortical connections in the developing cerebral cortex. Mutations in the gene are associate with cleft palate isolated (CPI).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SATB2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL0276
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SATB2
Gene Accession No. Q9UPW6
Gene Alias DNA-binding protein SATB2; FLJ21474; FLJ32076; GLSS; KIAA1034; KIAA1034-like DNA binding protein; MGC119474; MGC119477; mKIAA1034; RGD1562369; SATB family member 2; SATB homeobox 2; SATB2; special AT-rich sequence binding protein 2; special AT-rich sequence-binding protein 2; two cut domains-containing homeodomain protein
Gene Symbols SATB2
Host Species Mouse
Immunogen Recombinant protein fragment of Human SATB2 (Product #RP-93680)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23314
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.