Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Satumomab Chimeric Recombinant Rabbit Monoclonal Antibody (B72.3 (Satumomab))
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547862
Description
Specificity: Recognises human Tumor associated glycoprotein 72.
Satumomab is a biosimilar that targets TAG-72.
Specifications
Satumomab Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
Rabbit | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG κ |
Flow Cytometry, Immunohistochemistry, Immunomicroscopy | |
B72.3 (Satumomab) | |
Unconjugated | |
Indium (111In) satumomab pendetide; OncoScint CR; OncoScint CR103 | |
human Tumor associated glycoprotein 72. | |
200 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction