Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAV1 Antibody (OTI2B7), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273998AF532
Description
SAV1 Monoclonal specifically detects SAV1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SAV1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
SAV1 | |
Recombinant protein fragment corresponding to amino acids 167-383 of human SAV1 (NP_068590) produced in E.coli. | |
0.1 mL | |
60485.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2B7 | |
Western Blot | |
45 kDa WW domain protein, hWW45, protein salvador homolog 1, salvador homolog 1 (Drosophila), SAV, WW domain-containing, WW45salvador, WWP4,1700040G09Rik | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction