Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SAV1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597317 Supplier Invitrogen™ Supplier No. PA597317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPLPPGWSVD WTMRGRKYYI DHNTNTTHWS HPLEREGLPP GWERVESSEF GTYYVDHTNK KAQYRHPCAP SVPRYDQPPP VTYQPQQTER NQSLLVPANP YHTAEIPDWL QVYARAPVKY DHILKWELFQ LADLDTYQGM LKLLFMKELE QIVKMYEAYR QALLTELENR KQRQQWYAQQ HGKNF Positive Samples: HeLa, SKOV3, SW480; Cellular Location: Cytoplasm, Nucleus.

Sav1 is a regulator of STK3/MST2 and STK4/MST1 in the Hippo signaling pathway which plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 with its regulatory protein MOB1, which in turn phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. Phosphorylation of YAP1 by LATS1/2 inhibits its translocation into the nucleus to regulate cellular genes important for cell proliferation, cell death, and cell migration. SAV1 is required for STK3/MST2 and STK4/MST1 activation and promotes cell-cycle exit and terminal differentiation in developing epithelial tissues. It also plays a role in centrosome disjunction by regulating the localization of NEK2 to centrosomes, and its ability to phosphorylate CROCC and CEP250. In conjunction with STK3/MST2, Sav1 activates the transcriptional activity of ESR1 through the modulation of its phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAV1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene SAV1
Gene Accession No. Q8VEB2, Q9H4B6
Gene Alias 1700040G09Rik; 45 kDa WW domain protein; EMBL:CAJ98868.1}; hWW45; mWW45; Protein salvador homolog 1; rWW45; Salv; salvador family WW domain containing protein 1; salvador homolog 1; salvador homolog 1 (Drosophila); Salvador homolog 1 protein; salvador-like protein 1-like protein; Sav; Sav1; sav1 {ECO:0000312; WW domain-containing adaptor 45; WW domain-containing protein 3; WW domain-containing protein 4; WW45; WW45 protein; Wwp3; Wwp4
Gene Symbols SAV1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 199-383 of human SAV1 (NP_0685901).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60485, 64010
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.