Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SAV1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598928
Description
SAV1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA
Sav1 is a regulator of STK3/MST2 and STK4/MST1 in the Hippo signaling pathway which plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 with its regulatory protein MOB1, which in turn phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. Phosphorylation of YAP1 by LATS1/2 inhibits its translocation into the nucleus to regulate cellular genes important for cell proliferation, cell death, and cell migration. SAV1 is required for STK3/MST2 and STK4/MST1 activation and promotes cell-cycle exit and terminal differentiation in developing epithelial tissues. It also plays a role in centrosome disjunction by regulating the localization of NEK2 to centrosomes, and its ability to phosphorylate CROCC and CEP250. In conjunction with STK3/MST2, Sav1 activates the transcriptional activity of ESR1 through the modulation of its phosphorylation.
Specifications
SAV1 | |
Polyclonal | |
Unconjugated | |
SAV1 | |
1700040G09Rik; 45 kDa WW domain protein; EMBL:CAJ98868.1}; hWW45; mWW45; Protein salvador homolog 1; rWW45; Salv; salvador family WW domain containing protein 1; salvador homolog 1; salvador homolog 1 (Drosophila); Salvador homolog 1 protein; salvador-like protein 1-like protein; Sav; Sav1; sav1 {ECO:0000312; WW domain-containing adaptor 45; WW domain-containing protein 3; WW domain-containing protein 4; WW45; WW45 protein; Wwp3; Wwp4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
60485, 64010 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VEB2, Q9H4B6 | |
SAV1 | |
Recombinant Human Protein salvador homolog 1 protein (1-210AA). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction