Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Savant™ SpeedVac™ SPD120 Vacuum Concentrator and Kits
GSA_VA

Catalog No. p-7115393
Change view
Click to view available options
Description:
SpeedVac SPD120 Vacuum Concentrator
SpeedVac SPD120 Vacuum Concentrator
SpeedVac SPD120 Vacuum Concentrator Kit
Electrical Requirements:
115 V 60 Hz
230 V 50 Hz
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Electrical Requirements
13-875-341 SpeedVac SPD120 Vacuum Concentrator 115 V 60 Hz
13-875-342 SpeedVac SPD120 Vacuum Concentrator 230 V 50 Hz
13-875-363 SpeedVac SPD120 Vacuum Concentrator Kit 115 V 60 Hz
13-875-364 SpeedVac SPD120 Vacuum Concentrator Kit 230 V 50 Hz
13-875-365 SpeedVac SPD120 Vacuum Concentrator Kit 115 V 60 Hz
13-875-366 SpeedVac SPD120 Vacuum Concentrator Kit 230 V 50 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 13-875-341 Supplier Thermo Scientific™ Supplier No. SPD120115
Only null left
Add to Cart
Add to Cart
Request a Quote
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Obtain the necessary components for molecular biology applications with Thermo Scientific™ Savant™ SpeedVac™ SPD120 vacuum concentrator and kits.

These modular vacuum concentrators provide flexibility for your benchtop space. They are ideal for evaporation of non-aggressive organics. Choose from one of 2 kits available: SPD120P1 SpeedVac kit for standard evaporation or SPD120 P2 SpeedVac™ kit for lyophilization. All SpeedVac kits contain all the necessary components under one model number.

Space-saving design:

  • Automatic bleeder valve for un-attended start/stop
  • Digital display of parameters for easy, reproducible selection
  • Three user defined programs that can be modified and saved
  • Independent timers for both heat and run and audible alarm when run is completed
  • PTFE-coated chamber for superior chemical resistance and easy cleaning
  • Match up with an appropriate cold trap, vacuum pump or chemical trap for a complete concentration/drying system

Product Features:

  • Basic molecular biology applications with increased microplate capacity
  • Simple display and programming
  • Six programs available
  • Enhanced evaporation model, organic compatible
  • Radiant glass cover to accelerate evaporation and reduce drying time
  • Vacuum end point mode terminates run when preset vacuum level is reached
  • Digital display of vacuum, temperature and time section
  • Latched lid and lid lock indicator ensuring user safety during operation

SpeedVac SPD120 P1 Kit includes:

  • SpeedVac concentrator
  • Universal vacuum system (UVS450) including an oil-free vacuum pump and a -50 °C refrigerated vapor trap
  • Glass condensation flask (GCF400)
  • 40 x 1.5 mL rotor (RH40-11)
  • Tubing package including 12’ of thick-walled vacuum tubing, cutter, clamps and fittings, BV130 manual bleed valve and instructions
  • CryoCool heat transfer fluid (1 L)

SpeedVac SPD120 P2 Kit includes:

  • SpeedVac concentrator, basic molecular biology
  • -105 ºC, ultra-low temperature refrigerated vapor trap
  • VPL120 high vacuum pump
  • 40 x 1.5 mL rotor (RH40-11)
  • Universal tubing package (TFK100)
  • Digital vacuum gauge
  • Chemical trap kit for use with disposable cartridge
  • Disposable cartridge with color indicator for trapping acids and water vapor
  • Glass condensation flask
  • Vacuum pump lubricating oil
  • CryoCool™ heat transfer fluid (1 L)

Solvent and solvent combinations

  • Acetonitrile
  • Ethanol
  • Isopropanol
  • Methanol
  • Water
  • Acetonitrile and water
  • Ethanol and water
  • Methanol and water
  • Ammonium hydroxide

Typical applications:

  •  Drying biomolecules in HPLC fractions
  • Evaporating purification solvents
  • Drying extracts of natural products in drug assays
  • Lyophillization

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Model Medium capacity glass lid
Certifications UL Listed to US and Canadian requirements. CE marked
Description SpeedVac SPD120 Vacuum Concentrator
Electrical Requirements 115 V 60 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.