Learn More
Thermo Scientific™ Savant™ SpeedVac™ SPD300 : Large Capacity Vacuum Concentrator and Kits
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
Supplier: Thermo Scientific™ SPD300P1115
Description
Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.
- For strong acids and bases used in combinatorial chemistry
Specifications
| UL Listed to US and Canadian requirements. CE marked | |
| SpeedVac SPD300DDA Vacuum Concentrator Kit | |
| 115 V 60 Hz |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.