Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium diisopropylamide mono(tetrahydrofuran), 1.5M solution in cyclohexane, AcroSeal™, Thermo Scientific™

Catalog No. AC432878000
Click to view available options
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123333-84-6, 110-82-7
Molecular Formula C10H22LiNO
Molecular Weight (g/mol) 179.23
InChI Key YTJXGDYAEOTOCG-UHFFFAOYSA-N
Synonym lithium diisopropylamide mono tetrahydrofuran, lithium diisopropylamide mono tetrahydrofuran complex solution, lithium;di propan-2-yl azanide;oxolane, acmc-20ehmk, lda thf, lda.thf, lithiumdiisopropylamide thf, lithium diisopropylamide thf, lithiumdiisopropylamide tetrahydrofuran, lithium diisopropylamide tetrahydorfuran
PubChem CID 10997623
IUPAC Name lithium;di(propan-2-yl)azanide;oxolane
SMILES [Li+].CC(C)[N-]C(C)C.C1CCOC1

Specifications

Color Brown to Orange-Yellow
Physical Form Liquid
Quantity 800 mL
Flash Point −18°C
Formula Weight 179.23
Concentration or Composition (by Analyte or Components) 1.43 to 1.65M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.816
Density 0.8160g/mL
Chemical Name or Material Lithium diisopropylamide mono(tetrahydrofuran)
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.