Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saxagliptin

Catalog No. AC462180010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462180010 1 g
AC462182500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462180010 Supplier Thermo Scientific Chemicals Supplier No. 462180010
Add to Cart

CAS: 361442-04-8 | C18H25N3O2 | 315.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 361442-04-8
Molecular Formula C18H25N3O2
Molecular Weight (g/mol) 315.42
MDL Number MFCD12756398
InChI Key QGJUIPDUBHWZPV-UHFFFAOYNA-N
Synonym saxagliptin
PubChem CID 129317852
IUPAC Name (1R,5R)-2-[2-amino-2-[(7R)-3-hydroxy-1-adamantyl]acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile
SMILES NC(C(=O)N1C2CC2CC1C#N)C12CC3CC(CC(O)(C3)C1)C2

Specifications

Melting Point 107.0°C to 110.0°C
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in dmso, methanol
Formula Weight 315.41
Chemical Name or Material Saxagliptin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.