Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAYSD1 Antibody (OTI2H3), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272022APC
Description
SAYSD1 Monoclonal specifically detects SAYSD1 in Human samples. It is validated for Flow Cytometry.Specifications
| SAYSD1 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| SAYSD1 | |
| Full length recombinant protein of human C6orf64 (NP_060792) produced in HEK293T cells. | |
| 0.1 mL | |
| 55776.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2H3 | |
| Flow Cytometry | |
| C6orf64, chromosome 6 open reading frame 64, DKFZp434H012, FLJ11101, hypothetical protein LOC55776, SAYSVFN motif domain containing 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction