Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAYSD1 Antibody (OTI2H3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272022
Description
SAYSD1 Monoclonal specifically detects SAYSD1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
C6orf64 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100, CyTOF-ready | |
C6orf64, chromosome 6 open reading frame 64, DKFZp434H012, FLJ11101, hypothetical protein LOC55776, SAYSVFN motif domain containing 1 | |
Mouse | |
20 kDa | |
100 μg | |
55776 | |
Human | |
Lyophilized |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
OTI2H3 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
SAYSD1 | |
Full length human recombinant protein of human C6orf64(NP_060792) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction