Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAYSD1 Antibody (OTI2H3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272022H
Description
SAYSD1 Monoclonal specifically detects SAYSD1 in Human samples. It is validated for Western Blot.Specifications
SAYSD1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SAYSD1 | |
Full length recombinant protein of human C6orf64 (NP_060792) produced in HEK293T cells. | |
0.1 mL | |
55776.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2H3 | |
Western Blot | |
C6orf64, chromosome 6 open reading frame 64, DKFZp434H012, FLJ11101, hypothetical protein LOC55776, SAYSVFN motif domain containing 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction