Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Sazetidine A dihydrochloride
GREENER_CHOICESDP

Catalog No. 273610 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-361-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 27-361-0 Supplier R&D Systems™ Supplier No. 2736/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. α4β2 receptor ligand; may act as an agonist or a desensitizer

Subtype-selective α4β2 nicotinic acetylcholine receptor ligand (Ki values are 0.26 and 54 nM at α4β2 and α3β4 receptors respectively). May act as a silent desensitizer or as an agonist, depending on subunit stoichiometry (EC50 = 1.1 nM for nAChR-stimulated dopamine release). Exhibits analgesic activity in vivo and significantly reduces nicotine self-administration in an experimental rat model.

Chemical Identifiers

CAS 2455450-63-0
Molecular Formula C15H22Cl2N2O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 333.25
Purity >98%
Molecular Weight (g/mol) 333.25
Chemical Name or Material Sazetidine A dihydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.