Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SB 202190
GSA_VAGREENER_CHOICESDP

Catalog No. 126450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-645-0 50 mg
12-641-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-645-0 Supplier R&D Systems™ Supplier No. 1264/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective inhibitor of p38 MAPK

SB 202190 is a highly selective, potent and cell-permeable inhibitor of p38 MAP kinase. Binds within the ATP pocket of the active kinase (Kd = 38 nM, as measured in recombinant human p38), and selectively inhibits the p38α and β isoforms (IC50 = 50 and 100 nM at SAPK2a/p38 and SAPK2b/p38β2 respectively). Promotes stability of naive human pluripotent stem cells in culture. Inhibits Yamanaka factor reprogramming of human fibroblasts to iPSCs. SB 202190 improves the self-renewal ability of neuronal stem cells from NPC1-deficient mice.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 152121-30-7
Molecular Formula C20H14FN3O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 331.35
Purity >99%
Molecular Weight (g/mol) 331.35
Chemical Name or Material SB 202190
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.