Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SB 203580 hydrochloride
GREENER_CHOICESDP

Catalog No. 140210 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-021-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-021-0 Supplier R&D Systems™ Supplier No. 1402/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of p38 MAPK; water-soluble

Water-soluble salt of SB 203580 (Cat. No. 1202). Selective inhibitor of p38 mitogen-activated protein kinase (IC50 values are 50 and 500 nM for SAPK2a/p38 and SAPK2b/p38β2 respectively). Displays 100-500-fold selectivity over LCK, GSK3β and PKBα. Shown to inhibit interleukin-2-induced T cell proliferation, cyclooxygenase-1 and -2, and thromboxane synthase.

Chemical Identifiers

CAS 869185-85-3
Molecular Formula C21H17ClFN3OS

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 413.89
Purity >98%
Molecular Weight (g/mol) 413.90
Chemical Name or Material SB 203580 hydrochloride
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.