Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SB 218078
GREENER_CHOICESDP

Catalog No. 256010 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-601-0 10 mg
25-601 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-601-0 Supplier R&D Systems™ Supplier No. 2560/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Chk1 inhibitor

Inhibitor of checkpoint kinase 1 (Chk1) that displays selectivity over other protein kinases (IC50 values are 15, 250 and 1000 nM for Chk1, cdc2 and PKC respectively). Abrogates G2 cell cycle arrest caused by γ-irradiation and topoisomerase I inhibition. Potentiates cytotoxicity of DNA-damaging drugs, enhancing the efficacy of some chemotherapeutics.

Chemical Identifiers

CAS 135897-06-2
Molecular Formula C24H15N3O3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 393.39
Purity >98%
Molecular Weight (g/mol) 393.402
Chemical Name or Material SB 218078
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.