Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 297006
GREENER_CHOICE

Catalog No. 421310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-131-0 10 mg
42-135-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-131-0 Supplier Tocris Bioscience™ Supplier No. 4213/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective CCR3 antagonist

SB 297006 is a potent and selective CCR3 antagonist (IC50 = 39 nM). Displays 250-fold selectivity for CCR3 over other chemokine receptors, including CXCR1, CXCR2, CCR1 and CCR7 (IC50 >27 μM). Inhibits calcium mobilization induced by MCP-4, eotaxin-2 and eotaxin in RBL-2H3 cells transfected with CCR3 (IC50 values are 80, 90 and 210 nM respectively).

Specifications

Quantity 10 mg
Product Type SB 297006
Purity >99%
Molecular Weight (g/mol) 342.35
Inhibitors SB 297006

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.