Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 415286
GREENER_CHOICE

Catalog No. 161710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-171-0 10 mg
16-175-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-171-0 Supplier Tocris Bioscience™ Supplier No. 1617/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective GSK-3 inhibitor

Potent and selective glycogen synthase kinase-3 (GSK-3) inhibitor (Ki = 31 nM for GSK-3α); competes with ATP. Has minimal activity against 24 other protein kinases (IC50 > 10 μ M). Stimulates glycogen synthesis, gene transcription and is neuroprotective.

Chemical Identifiers

CAS 264218-23-7
Molecular Formula C16H10ClN3O5

Specifications

Quantity 10 mg
Formula Weight 359.73
Purity >99%
Molecular Weight (g/mol) 359.722
Chemical Name or Material SB 415286
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.