Learn More
Description
For further information on how SB 431542 may be used please see our protocol: Accelerated Induction of Cortical Neurons from hiPSCs .
Research use only SB 431542 also available.
Specifications
Specifications
| Chemical Name or Material | 4-[4-(1,3-benzodioxol-5-yl)-5-(2-pyridinyl)-1H-imidazol-2-yl]benzamide |
| CAS | 301836-41-9 |
| Quantity | 10 mg |
| Target | TGF-beta Receptor Inhibitors |
| Molecular Formula | C22H16N4O3 |
| Purity | 99% |
| Regulatory Status | GMP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
