Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 525334
GREENER_CHOICE

Catalog No. 321110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-111-0 10 mg
32-115-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-111-0 Supplier Tocris Bioscience™ Supplier No. 3211/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective inhibitor of TGF-βRI

SB 525334 is a selective inhibitor of transforming growth factor-β receptor I (ALK5, TGF-βRI) (IC50 = 14.3 nM). Inhibits TGF-β1-induced smad2/3 nuclear localization and TGF-βRI-induced mRNA expression in kidney cells. Attenuates bleomycin-induced pulmonary fibrosis.

Specifications

Quantity 10 mg
Product Type SB 525334
Purity >97%
Molecular Weight (g/mol) 343.42
Inhibitors SB 525334

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.