Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SB 590885
GREENER_CHOICESDP

Catalog No. 265010 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-501-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-501-0 Supplier R&D Systems™ Supplier No. 2650/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent B-Raf inhibitor

Potent B-Raf inhibitor (Kd = 0.3 nM). Selective for B-Raf against 46 other kinases (Ki app values are 0.16 and 1.72 nM for B-Raf and c-Raf respectively). Decreases anchorage-independent growth of melanoma cell lines. Inhibits ERK phosphorylation and proliferation in tumor cells expressing B-RafV600E.

Chemical Identifiers

CAS 405554-55-4
Molecular Formula C27H27N5O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 453.54
Purity >98%
Molecular Weight (g/mol) 453.55
Chemical Name or Material SB 590885
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.