Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 612111 hydrochloride
GREENER_CHOICE

Catalog No. 357310R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-731-0R 10 mg
35-735-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-731-0R Supplier Tocris Bioscience™ Supplier No. 3573/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective NOP antagonist

SB 612111 hydrochloride is a selective NOP receptor antagonist (Ki values are 0.33, 57.6, 160.5 and 2109 nM for NOP, μ-, κ- and δ-receptors respectively). Antagonizes the pronociceptive action of nociceptin (Cat. No. 0910) in an acute pain model. Potentiates the action of morphine in morphine-tolerant animals and blocks hyperalgesia in an inflammatory pain model.

Specifications

Quantity 10 mg
Molecular Formula (5 S,7 S)-7-[[4-(2,6-Dichlorophenyl)-1-piperidinyl]methyl]-6,7,8,9-tetrahydro-1-methyl-5 H-benzocyclohepten-5-ol hydrochloride
Product Type SB 612111 hydrochloride
Purity 97%
Molecular Weight (g/mol) M.W. (Observed): 454.86
Inhibitors SB 612111 hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.