Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 657510
SDP

Catalog No. p-5546015 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-715-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 35-715-0R Supplier Tocris Bioscience™ Supplier No. 3571/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective urotensin-II (UT) receptor antagonist

Selective urotensin-II (UT) receptor antagonist (K i values are 61, 17, 30, 65 and 56 nM at human, monkey, cat, rat and mouse receptors respectively). Inhibits U-II-induced intracellular Ca 2+ mobilization (IC 50 = 180 nM) and antagonizes the contractile action of U-II in isolated mammalian arteries and aortae (EC 50 = 50 - 189 nM).

Specifications

Purity >98%
Quantity 50 mg
Product Type SB 657510
Molecular Weight (g/mol) 505.81
Inhibitors SB 657510

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.