Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SB 674042
GREENER_CHOICE

Catalog No. 467310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-731-0 10 mg
46-735-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-731-0 Supplier Tocris Bioscience™ Supplier No. 4673/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective non-peptide OX1 antagonist

Potent and selective non-peptide orexin OX1 receptor antagonist (Kd = 3.76 nM). Exhibits 100-fold selectivity for OX1 over OX2 receptors. Has no effect at serotonergic, dopaminergic, adrenergic or purinergic receptors. Inhibits orexin 1-induced Ca2+ mobilization in CHO-DG44 cells stably transfected with the OX1 receptor.

Chemical Identifiers

CAS 483313-22-0
Molecular Formula C24H21FN4O2S

Specifications

Quantity 10 mg
Formula Weight 448.51
Molecular Weight (g/mol) 448.516
Chemical Name or Material SB 674042
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.