Learn More
Invitrogen™ SBCAD Polyclonal Antibody

Description
Immunogen sequence: EASMAKYYAS EIAGQTTSKC IEWMGGVGYT KDYPVEKYFR DAKIGTIYEG ASNIQLNTIA KHIDAE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 92%.
Specifications
Specifications
| Antigen | SBCAD |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ACADSB |
| Gene Accession No. | P45954 |
| Gene Alias | 1300003O09Rik; 2-MEBCAD; 2-methyl branched chain acyl-CoA dehydrogenase; 2-methylbutyryl-CoA dehydrogenase; 2-methylbutyryl-coenzyme A dehydrogenase; ACAD7; Acadsb; acyl-CoA dehydrogenase short/branched chain; acyl-CoA dehydrogenase, short/branched chain; Acyl-Coenzyme A dehydrogenase short-branched chain; acyl-Coenzyme A dehydrogenase, short/branched chain; BB066609; SBCAD; Short/branched chain specific acyl-CoA dehydrogenase, mitochondrial |
| Gene Symbols | ACADSB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.