Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SBCAD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112303
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112303 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112303 Supplier Invitrogen™ Supplier No. PA5112303
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SBCAD Polyclonal Antibody for Western Blot, IHC (P), IP

Short/branched chain acyl-CoA dehydrogenase(ACADSB) is a member of the acyl-CoA dehydrogenase family of enzymes that catalyze the dehydrogenation of acyl-CoA derivatives in the metabolism of fatty acids or branch chained amino acids. Substrate specificity is the primary characteristic used to define members of this gene family. The ACADSB gene product has the greatest activity towards the short branched chain acyl-CoA derivative, (S)-2-methylbutyryl-CoA, but also reacts significantly with other 2-methyl branched chain substrates and with short straight chain acyl-CoAs. The cDNA encodes for a mitochondrial precursor protein which is cleaved upon mitochondrial import and predicted to yield a mature peptide of approximately 43.7- kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SBCAD
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene ACADSB
Gene Accession No. P45954
Gene Alias 1300003O09Rik; 2-MEBCAD; 2-methyl branched chain acyl-CoA dehydrogenase; 2-methylbutyryl-CoA dehydrogenase; 2-methylbutyryl-coenzyme A dehydrogenase; ACAD7; Acadsb; acyl-CoA dehydrogenase short/branched chain; acyl-CoA dehydrogenase, short/branched chain; Acyl-Coenzyme A dehydrogenase short-branched chain; acyl-Coenzyme A dehydrogenase, short/branched chain; BB066609; SBCAD; Short/branched chain specific acyl-CoA dehydrogenase, mitochondrial
Gene Symbols ACADSB
Host Species Rabbit
Immunogen E. coli-derived Human ACADSB fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 36
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.