Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SBDS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528950
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528950 Supplier Invitrogen™ Supplier No. PA528950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse liver. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (82%), Xenopus laevis (86%), Pig (99%), Chicken (89%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of a highly conserved protein family that exists from archaea to vertebrates and plants. The encoded protein may function in RNA metabolism. Mutations within this gene are associated with Shwachman-Bodian-Diamond syndrome. An alternative transcript has been described, but its biological nature has not been determined. This gene has a closely linked pseudogene that is distally located.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SBDS
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SBDS
Gene Accession No. P70122, Q9Y3A5
Gene Alias 4733401P19Rik; AI836084; CGI-97; FLJ10917; Protein 22A3; ribosome maturation protein SBDS; SBDS; SBDS ribosome assembly guanine nucleotide exchange factor; SBDS ribosome maturation factor; SBDS, ribosome assembly guanine nucleotide exchange factor; SBDS, ribosome maturation factor; SDS; Shwachman-Bodian-Diamond syndrome; Shwachman-Bodian-Diamond syndrome homolog; Shwachman-Bodian-Diamond syndrome homolog (human); Shwachman-Bodian-Diamond syndrome protein; Shwachman-Bodian-Diamond syndrome protein homolog; SWDS; zgc:56700
Gene Symbols SBDS
Host Species Rabbit
Immunogen Full length human SBDS Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51119, 66711
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.