Learn More
SBDS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 176181AP150UL

Description
This antibody is a rabbit polyclonal antibody raised against a full-length human SBDS protein, recognizes specifically the 29 kDa SBDS protein.
This gene encodes a member of a highly conserved protein family that exists from archaea to vertebrates and plants. The encoded protein may function in RNA metabolism. Mutations within this gene are associated with Shwachman-Bodian-Diamond syndrome. An alternative transcript has been described, but its biological nature has not been determined. This gene has a closely linked pseudogene that is distally located.Specifications
SBDS | |
Polyclonal | |
Unconjugated | |
SBDS | |
CGI 97, SBDS, SDS, SWDS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
288615, 51119, 66711 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70122, Q5RK30, Q9Y3A5 | |
Sbds | |
SBDS Fusion Protein Ag11814 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.