Learn More
Invitrogen™ SBDS Recombinant Rabbit Monoclonal Antibody (9V4J9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542626
Description
Positive test controls include: TE-1, HepG2, SKOV3, Mouse lung, Mouse liver, Mouse brain, Mouse heart, Rat lung, Rat kidney. The target is usually found in the following locations: Cytoplasm, Nucleus, cytoskeleton, nucleolus, nucleoplasm, spindle. Immunogen sequence: KQQALEVIKQ LKEKMKIERA HMRLRFILPV NEGKKLKEKL KPLIKVIESE DYGQQLEIVC LIDPGCFREI DELIKKETKG KGSLEVLNLK DVEEGDEKFE.
This gene encodes a member of a highly conserved protein family that exists from archaea to vertebrates and plants. The encoded protein may function in RNA metabolism. Mutations within this gene are associated with Shwachman-Bodian-Diamond syndrome. An alternative transcript has been described, but its biological nature has not been determined. This gene has a closely linked pseudogene that is distally located.
Specifications
SBDS | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P70122, Q5RK30, Q9Y3A5 | |
SBDS | |
A synthetic peptide corresponding to a sequence within amino acids 151-250 of human SBDS (NP_0571222). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
9V4J9 | |
Unconjugated | |
SBDS | |
4733401P19Rik; AI836084; CGI-97; FLJ10917; Protein 22A3; ribosome maturation protein SBDS; SBDS; SBDS ribosome assembly guanine nucleotide exchange factor; SBDS ribosome maturation factor; SBDS, ribosome assembly guanine nucleotide exchange factor; SBDS, ribosome maturation factor; SDS; Shwachman-Bodian-Diamond syndrome; Shwachman-Bodian-Diamond syndrome homolog; Shwachman-Bodian-Diamond syndrome homolog (human); Shwachman-Bodian-Diamond syndrome protein; Shwachman-Bodian-Diamond syndrome protein homolog; SWDS; zgc:56700 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
288615, 51119, 66711 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.