Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SBEM Antibody (Mc5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281060F
Description
SBEM Monoclonal specifically detects SBEM in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
SBEM | |
Monoclonal | |
FITC | |
PBS | |
MUCL1 | |
Human SBEM. | |
0.1 mL | |
Primary | |
Specifically binds SBEM. | |
Store at 4°C in the dark. | |
IgG2a κ |
ELISA, Immunocytochemistry, Immunofluorescence | |
Mc5 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
mucin-like 1, mucin-like protein 1, Protein BS106, SBEMSmall breast epithelial mucin | |
Mouse | |
Protein A purified | |
RUO | |
118430 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction