Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SBK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | SBK1 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17414820
![]() |
Novus Biologicals
NBP17414820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP174148
![]() |
Novus Biologicals
NBP174148 |
100 μL |
Each for $487.50
|
|
|||||
Description
SBK1 Polyclonal specifically detects SBK1 in Mouse samples. It is validated for Western Blot.Specifications
SBK1 | |
Western Blot | |
Unconjugated | |
RUO | |
EC 2.7.11, EC 2.7.11.1, Sbk, serine/threonine-protein kinase SBK1, SH3-binding domain kinase 1, SH3-binding kinase 1 | |
SBK1 | |
IgG |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Rabbit | |
Q8QZX0 | |
388228 | |
Synthetic peptides corresponding to the C terminal of Sbk1. Immunizing peptide sequence GSRPSPPSVGPVVPVPVPVPVPVPEAGLAPPAPPGRTDGRTDKSKGQVVL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title