Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Sbk1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549384
Description
Predicted to react with rat based on sequence homology.
SBK1 novel serine/threonine kinase or SH3 domain binding kinase 1 is structurally related to a Xenopus gastrula-specific protein kinase, Pk9.7 which possessing a consensus sequence for an SH3-binding domain from developing rat brain. SBK1 plays an important role in signal-transduction pathways related to the control of brain development. SBK1 may predict a broad cellular function, and its dysregulated in certain cancers suggests an involvement of the protein in the pathogenesis of human cancers. May be involved in signal-transduction pathways related to the control of brain development. Target protein is a serine/threonine protein kinase gene expressed predominantly in developing brain.
Specifications
Sbk1 | |
Polyclonal | |
Unconjugated | |
SBK1 | |
SBK; SBK1; Serine/threonine-protein kinase SBK1; SH3 domain binding kinase 1; SH3-binding domain kinase 1; SH3-binding kinase 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
104175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8QZX0 | |
SBK1 | |
KLH conjugated synthetic peptide between 65-98 amino acids from the N-terminal region of Mouse Sbk1. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction