Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sbk2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556526
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556526 Supplier Invitrogen™ Supplier No. PA556526
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PENVLVCDPA CRRFKLTDFG HTRPRGTLLR LAGPPIPYTA PELCAPPPLP EGLPIQPALD AWALGVLLFC LLTGYFPWDR PLAEADPFYE DFLIWQASGQ PRDRPQPWFG LAPAADALLR GLLDPHPRRR SAVIAIWEHL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Sbk2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SBK2
Gene Accession No. P0C263
Gene Alias Gm1079; Sbk2; Serine/threonine-protein kinase SBK2; SGK069; SH3 domain binding kinase family member 2; SH3 domain binding kinase family, member 2; SH3-binding domain kinase family member 2; SH3-binding domain kinase family, member 2; Sugen kinase 69
Gene Symbols SBK2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Sbk2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 646643
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.