Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SbmA Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5143802
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5143802 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5143802 Supplier Invitrogen™ Supplier No. PA5143802
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This polyclonal antibody recognizes the inner membrane protein SbmA of Gram-negative bacteria.

SbmA is a gene that encodes a protein called inner membrane transporter SbmA, which is involved in the transport of various molecules across the inner membrane of bacterial cells. SbmA is particularly important for the uptake of peptide antibiotics, such as bacitracin and gramicidin, which are used in the treatment of bacterial infections. SbmA is also involved in the regulation of bacterial cell division and morphology. Understanding the function and regulation of SbmA is important for developing new antibiotics and for gaining insights into the mechanisms of bacterial resistance to antibiotics.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SbmA
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Host Species Rabbit
Immunogen Recombinant SbmA.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype Ig
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.