Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SBNO1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305794AF647
Description
SBNO1 Polyclonal specifically detects SBNO1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SBNO1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
FLJ10701, FLJ10833, FLJ14066, FLJ16176, FLJ35199, monocyte protein 3, MOP3, protein strawberry notch homolog 1, Sno, strawberry notch homolog 1 (Drosophila) | |
Produced in rabbits immunized with E. coli-derived Human SBNO1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55206 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction