Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560672 Supplier Invitrogen™ Supplier No. PA560672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEGIKLSADV KPFVPRFAGL NVAWLESSEA CVFPSSAATY YPFVQEPPVT EQKIYTEDMA FGASTFPPQY LSSEITLHPY AYSPYTLDST QNVYSVPGSQ YLYNQPSCYR GFQTVKHRNE NT Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.

Eukaryotes require a selenocysteine (Sec) insertion sequence (SECIS) element in the 3' untranslated region of the mRNA to decode the UGA codon as Sec. SECIS-binding protein 2 (SBP-2) specifically binds selenoprotein mRNAs to form a functional complex and is essential for the insertion of Sec into selenoproteins. Purified SBP-2 interacts specifically with the SECIS element in the phospholipid hydroperoxide glutathione peroxidase mRNA. SBP-2 shows binding activity in the liver and testis as well as hepatoma cells. SBP-2 binds to a conserved RNA binding domain shared with several ribosomal proteins and eukaryotic translation termination release factor 1. A second domain located N-terminal to the RNA binding domain required for Sec insertion allows SBP-2 to stably associate with the ribosomal fraction of cells. SBP-2 preferentially stimulates incorporation directed by the selenoprotein P and phospholipid hydroperoxide glutathione peroxidase SECIS elements. SBP-2 may have a distinct function in selecting the ribosomes for Sec insertion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SBP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SECISBP2
Gene Accession No. Q96T21
Gene Alias 2210413N07Rik; 2810012K13Rik; DKFZp686C09169; OTTHUMP00000064929; OTTHUMP00000064930; OTTHUMP00000064931; OTTHUMP00000064932; RP11-89K14.1; Sbp2; SECIS binding protein 2; SECIS-binding protein 2; SECISBP2; Selenocysteine insertion sequence-binding protein 2
Gene Symbols SECISBP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SBP2. Recombinant protein control fragment (Product #RP-93081).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79048
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.