Learn More
SBPP Level 1 Isolation Gowns
SDP
Catalog No. usbp1025cf Shop All Fisherbrand Products
Click to view available options
:
One Size
This item is not returnable. View return policy
This item is not returnable. View return policy
Provides basic protection from minor splashes and spill
  • Fabric: Non woven
  • Elastic cuff
  • Spunbonded polypropylene
  • Latex free
  • Lightweight and breathable
Specifications
Size Universal
Product Type Isolation Gown
Gender Unisex
No. of Pockets None
Material Polypropylene
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Latex Free Yes
Autoclavable Not Autoclavable
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.