Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SBSN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564338
Description
Immunogen sequence: HQAGKEAEKL GQGVNHAADQ AGKEVEKLGQ GAHHAAGQAG KELQNAHNGV NQASKEANQL LNGNHQSGSS Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 76%.
SBSN is a protein coding gene.
Specifications
SBSN | |
Polyclonal | |
Unconjugated | |
SBSN | |
HLAR698; SBSN; Suprabasin; UNQ698; UNQ698/PRO1343 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
374897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWP8 | |
SBSN | |
Recombinant protein corresponding to Human SBSN. Recombinant protein control fragment (Product #RP-103932). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction